Assay Details
| Gene Symbol: Fam179b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK132890.1 | - | 1 - 2 | 76 | 95 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | kelch-like 28 | Klhl28 |
2810440N09Rik, 4122402F11Rik, 4931401E10Rik, Btbd5 |
-15.0kb | Sense | Chr. 12 - 64942198 - 64965605 on NCBI Build GRCm38 |
| Downstream | PRP39 pre-mRNA processing factor 39 homolog (yeast) | Prpf39 | Srcs1 | 53.4kb | Anti-sense | Chr. 12 - 65036309 - 65063386 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Fam179b
Genomic Map