Assay Details
| Gene Symbol: 1700092E19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045933.1 | 1 - 2 | 284 | 90 | ||
| GenBank mRNA | AK133453.1 | - | 1 - 2 | 288 | 90 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 11351 | Gm11351 | OTTMUSG00000000620 | -319.8kb | Sense | Chr. 13 - 25921033 - 25966316 on NCBI Build GRCm38 |
| Downstream | hepatoma derived growth factor-like 1 | Hdgfl1 |
4921520N01Rik, HRP-1, Hdgfrp1, Pwwp1 |
455.8kb | Sense | Chr. 13 - 26768172 - 26770165 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC1700000603.rn
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2322234 | AK133453.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 304215 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 494809 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 192546 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 21758 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 149751 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 327991 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 61197 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 173565 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 347869 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 29739 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07816415 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07816457 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125781 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P07816457 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10478706 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897072 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897057 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897039 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897047 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10478710 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28896913 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897083 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28896844 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28896953 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897036 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897054 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897082 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897055 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28896841 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28896854 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28896848 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28896884 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897079 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897034 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897064 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10982495 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897080 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897065 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897006 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10982496 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28896852 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897081 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28896908 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28896830 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10757421 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28896910 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10589710 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10854710 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28896958 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28896914 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897017 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28896885 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28896917 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897089 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28896918 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897011 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10868486 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897048 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897029 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28896828 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28896850 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28896998 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28896849 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10868479 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897004 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897077 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28896952 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28896824 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897042 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28896896 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28896832 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897002 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897062 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28896889 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28896855 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897059 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897038 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28896840 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10982508 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897003 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897052 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897013 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897014 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897022 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897025 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28896837 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897031 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897069 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10982494 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10757386 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10537858 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897045 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28896826 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897090 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897060 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10788925 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897067 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28896901 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897030 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897000 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28896959 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897027 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897024 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897005 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897058 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10868487 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28896829 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28896856 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28896834 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897037 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897032 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10982506 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897076 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28896955 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897043 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897073 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28896996 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28896992 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10439641 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10868492 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28896962 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28896964 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28897012 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1457210_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1457210_at | - | - |
| Mouse Genome 430 2.0 Array | 1457210_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300010456.mm.1 | NR_045933.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300010456.mm.1 | NR_045933.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300010456.mm.1 | TC1300001680.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300010460.mm.1 | NR_045933.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300010460.mm.1 | NR_045933.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300010460.mm.1 | TC1300001680.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300010455.mm.1 | NR_045933.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300010455.mm.1 | NR_045933.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300010455.mm.1 | TC1300001680.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300010454.mm.1 | NR_045933.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300010454.mm.1 | NR_045933.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300010454.mm.1 | TC1300001680.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300010461.mm.1 | NR_045933.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300010461.mm.1 | NR_045933.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300010461.mm.1 | TC1300001680.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1457210_at | - | - |
| Mouse Expression Set 430 | 1457210_PM_at | - | - |
| Murine Genome U74v2 Set | 167015_PM_at | - | - |
| Murine Genome U74v2 Set | 167015_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1457210_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1457210_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 103710541 | - | - |

1700092E19Rik
Genomic Map