Assay Details
| Gene Symbol: Pax7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK133693.1 | - | 1 | 2250 | 101 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
taste receptor, type 1, member 2 |
Tas1r2 |
Gpr71, T1r2, TR2 |
-66.8kb | Anti-sense | Chr. 4 - 139653538 - 139670280 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 4933427I22 gene | 4933427I22Rik | 186.7kb | Sense | Chr. 4 - 139926111 - 139943519 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pax7
Genomic Map