Assay Details
| Gene Symbol: Gm1653 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040591.1 | 1 - 2 | 452 | 62 | ||
| GenBank mRNA | AK133793.1 | - | 1 - 2 | 453 | 62 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 36888 |
Gm36888 | -1.6mb | Anti-sense | Chr. 3 - 147620811 - 147635647 on NCBI Build GRCm38 | |
| Downstream | predicted gene 5149 | Gm5149 | EG381483 | 58.6kb | Sense | Chr. 3 - 149335731 - 149340637 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm1653
Genomic Map