Assay Details
| Gene Symbol: 5330413P13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK134994.1 | - | 1 - 2 | 88 | 76 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 14285 | Gm14285 | OTTMUSG00000016191 | -280.5kb | Anti-sense | Chr. 2 - 131561614 - 131563181 on NCBI Build GRCm38 |
| Downstream | endogenous retroviral sequence 3 | Erv3 |
1600014E20Rik, AA389086, AI426069, AI448024, D19349 |
6.0kb | Sense | Chr. 2 - 131853678 - 131889598 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

5330413P13Rik
Genomic Map