Assay Details
| Gene Symbol: Gm5817 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK135560.1 | - | 2 - 3 | 879 | 99 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
protein kinase C, epsilon |
Prkce |
5830406C15Rik, PKC[e], PKCepsilon, Pkce, R75156 |
Anti-sense | Chr. 17 - 86166971 - 86657919 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 2010106C02 gene | 2010106C02Rik | 118.1kb | Sense | Chr. 17 - 86285167 - 86287178 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm5817
Genomic Map