Assay Details
| Gene Symbol: Gm20759 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046030.1 | 1 - 2 | 160 | 74 | ||
| GenBank mRNA | AK135737.1 | - | 1 - 2 | 161 | 74 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | T-box 4 | Tbx4 | 3930401C23 | -106.3kb | Anti-sense | Chr. 11 - 85886343 - 85916099 on NCBI Build GRCm38 |
| Downstream | BRCA1 interacting protein C-terminal helicase 1 | Brip1 |
3110009N10Rik, 8030460J03Rik, Bach1, FACJ, Fancj, OF |
33.8kb | Sense | Chr. 11 - 86058131 - 86201267 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC1000003463.rn
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 686237 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 587897 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 112642 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 928241 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 819913 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 123363 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07427517 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986761 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09087924 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09541931 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09541935 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986808 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986751 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986748 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09006323 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986793 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09493994 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986679 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986757 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986690 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986635 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986588 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986584 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986790 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986782 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986618 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986687 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986664 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986662 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986638 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09355919 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986658 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986804 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986712 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986798 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986682 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986806 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986627 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986674 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986607 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986609 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986717 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986585 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986629 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986632 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986579 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986625 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986787 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986672 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986744 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986821 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986758 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986680 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09541927 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986693 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09261941 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986656 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986630 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986604 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09242835 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09242824 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986734 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09484417 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986659 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986599 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09033544 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986583 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986716 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986764 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986786 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986640 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986581 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09484413 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986780 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09333575 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986633 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986749 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986654 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986602 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09704983 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986719 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09493986 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986725 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986774 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09698469 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09325813 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986803 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986676 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986594 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986586 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986587 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986812 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986743 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986763 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986795 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986788 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986721 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09541930 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986681 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986760 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986791 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986714 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986649 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986634 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986598 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986811 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986731 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09493996 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986616 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986684 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09541932 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986739 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986753 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986816 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986582 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986685 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09086328 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986623 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986729 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986755 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986620 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09350085 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986802 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986817 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986741 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09541913 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986636 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986727 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09669494 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27986710 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Clariom S Array | PSR1100031492.mm.1 | NR_046030.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100031492.mm.1 | NR_046030.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100031492.mm.1 | TC1100003433.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100031488.mm.1 | NR_046030.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100031488.mm.1 | NR_046030.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100031488.mm.1 | TC1100003433.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100031490.mm.1 | NR_046030.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100031490.mm.1 | NR_046030.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100031490.mm.1 | TC1100003433.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100031489.mm.1 | NR_046030.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100031489.mm.1 | NR_046030.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100031489.mm.1 | TC1100003433.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100031491.mm.1 | NR_046030.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100031491.mm.1 | NR_046030.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100031491.mm.1 | TC1100003433.mm.1 | MTA TC |

Gm20759
Genomic Map