Assay Details
Gene Symbol: LOC101055656 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC086661.1 | - | 2 | 921 | 61 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | Meis homeobox 1 | Meis1 |
C530044H18Rik, Evi8 |
Anti-sense | Chr. 11 - 18880428 - 19020482 on NCBI Build GRCm38 | |
Downstream | RIKEN cDNA 4933406G16 gene | 4933406G16Rik | 54.1kb | Sense | Chr. 11 - 19065275 - 19075740 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
