Assay Details
| Gene Symbol: Gm626 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK136780.1 | - | 1 - 2 | 32 | 65 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mitogen-activated protein kinase 8 | Mapk8 |
AI849689, JNK, JNK1, Prkm8, SAPK1 |
-118.1kb | Anti-sense | Chr. 14 - 33377898 - 33447214 on NCBI Build GRCm38 |
| Downstream |
protein tyrosine phosphatase, non-receptor type 20 |
Ptpn20 | typ | 23.1kb | Sense | Chr. 14 - 33589201 - 33640754 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm626
Genomic Map