Assay Details
| Gene Symbol: Gm4593 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK138221.1 | - | 127 | 65 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | a disintegrin and metallopeptidase domain 12 (meltrin alpha) | Adam12 |
Mltna, mKIAA4001 |
-267.0kb | Sense | Chr. 7 - 133883196 - 134232149 on NCBI Build GRCm38 |
| Downstream | dedicator of cytokinesis 1 | Dock1 |
9130006G06Rik, AI854900, D630004B07Rik |
169.4kb | Anti-sense | Chr. 7 - 134670608 - 135173647 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm4593
Genomic Map