Assay Details
| Gene Symbol: Gm6209 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK138462.1 | - | 4 - 5 | 372 | 75 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
solute carrier family 7 (cationic amino acid transporter, y+ system), member 11 |
Slc7a11 |
9930009M05Rik, AI451155, sut, xCT |
-17.3kb | Sense | Chr. 3 - 50364936 - 50499087 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 37548 |
Gm37548 | 333.8kb | Sense | Chr. 3 - 50883237 - 50888892 on NCBI Build GRCm38 |
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2091039 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 100382 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 568742 | AK012924.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 28352 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 543250 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 511770 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 738419 | AK134427.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 738419 | BC019150.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 738419 | NM_013555.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 10001 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 833473 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 883068 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 549297 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 115023 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 518869 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 475050 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 382577 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 412661 | AK002620.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 412661 | AK028027.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 412661 | AK049398.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 412661 | AK083888.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 412661 | AK132029.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 412661 | AK145755.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 412661 | AK146305.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 412661 | AK146473.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 412661 | AK150750.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 412661 | AK151485.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 412661 | AK151564.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 412661 | AK151640.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 412661 | AK151687.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 412661 | AK152272.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 412661 | AK152317.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 412661 | BC043084.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 412661 | BC054811.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 412661 | D78188.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 412661 | NM_008098.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 167169 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 626514 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 232762 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 564315 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 451296 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 723384 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 837593 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 372004 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 89557 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 395149 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 858559 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 793131 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 809690 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 18938 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 365276 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 5898 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 33409 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 11545 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 9542 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 12420 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 130882 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2091039 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00638076 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04901835 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04901816 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04901715 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04901766 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04901883 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04901860 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04901738 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P04901715 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P00638076 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P04901883 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125345 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02420160 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03136599 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125360 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125053 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125307 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125051 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125355 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125316 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125059 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125269 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02791476 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125263 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125338 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125295 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125083 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125089 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125340 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02997914 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125352 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125365 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125289 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125359 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22124894 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125369 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22124895 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02695716 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02765903 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125275 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22124878 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125361 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125290 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125272 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125299 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125343 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02896251 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125044 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03110156 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22124885 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125305 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125344 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125043 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125082 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125261 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125342 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22124884 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22124891 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03119307 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125366 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125323 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125321 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22124900 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125318 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125293 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125335 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125086 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125052 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125054 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125062 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02588836 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02588827 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125320 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22124880 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125324 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125265 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125302 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125311 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125337 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125058 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125306 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125298 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125259 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125049 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125085 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125367 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125287 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125079 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125281 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125333 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03136595 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125055 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125363 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125270 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125078 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125285 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22124888 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125347 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125329 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125349 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125041 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125267 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125045 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125351 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125330 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125057 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125278 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02334174 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125231 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02344796 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125357 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03119294 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22124893 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125274 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22124877 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125277 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125090 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22124899 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02420146 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125047 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22124882 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125292 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22125075 | - | - |

Gm6209
Genomic Map