Assay Details
| Gene Symbol: Gm5547 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045845.1 | 1 - 2 | 25 | 64 | ||
| GenBank mRNA | AK138691.1 | - | 1 - 2 | 25 | 64 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RAS-related protein-1a | Rap1a |
AI848598, G-22K, Krev-1, Rap1 |
-14.2kb | Anti-sense | Chr. 3 - 105727260 - 105801381 on NCBI Build GRCm38 |
| Downstream | transmembrane and immunoglobulin domain containing 3 | Tmigd3 |
1700001d09rik, 4930578J19Rik |
54.9kb | Sense | Chr. 3 - 105870858 - 105924044 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm5547
Genomic Map