Assay Details
| Gene Symbol: B130034C11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK139053.1 | - | 1 - 2 | 170 | 95 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
chaperonin containing Tcp1, subunit 8 (theta) |
Cct8 |
AI132397, Cctq, Tcpq |
-130.0 | Anti-sense | Chr. 16 - 87483325 - 87495869 on NCBI Build GRCm38 |
| Downstream | Map3k7 C-terminal like | Map3k7cl |
C21orf7, ORF63, Tak1l |
49.5kb | Sense | Chr. 16 - 87553330 - 87595336 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

B130034C11Rik
Genomic Map