Assay Details
| Gene Symbol: Gm2155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK139105.1 | - | 382 | 65 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 449 | Zfp449 | 9530049C15Rik | -7.2kb | Anti-sense | Chr. X - 56346383 - 56365674 on NCBI Build GRCm38 |
| Downstream | DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 26B | Ddx26b |
4930535D10Rik, 6330505F04Rik, D130066O12 |
80.0kb | Anti-sense | Chr. X - 56454821 - 56507843 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm2155
Genomic Map