Assay Details
| Gene Symbol: D16Ertd519e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040474.1 | 2 - 3 | 206 | 95 | ||
| GenBank mRNA | AK139678.1 | - | 2 - 3 | 207 | 95 | |
| BG068156.2 | - | 1 - 2 | 95 | |||
| CO811502.1 | - | 2 - 3 | 215 | 95 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
glucan (1,4-alpha-), branching enzyme 1 |
Gbe1 |
2310045H19Rik, 2810426P10Rik, D16Ertd536e |
-51.4kb | Sense | Chr. 16 - 70313949 - 70569720 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 4931420L22 gene | 4931420L22Rik | 506.3kb | Sense | Chr. 16 - 71131076 - 71139029 on NCBI Build GRCm38 |
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P14856 | AK139678.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P14856 | AU022163.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P14856 | AK139678.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P14856 | AU022163.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 535226 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 257420 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 101046 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P14856 | AK139678.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P14856 | AU022163.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P14856 | AK139678.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P14856 | AU022163.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P14856 | AK139678.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P14856 | AU022163.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P14856 | AK139678.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P14856 | AU022163.1 | mouse_transcriptome |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851041 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851058 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851080 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30850966 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851125 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851118 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851047 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851092 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30850936 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30850956 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851105 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851038 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851102 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12635878 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12739313 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851128 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30850931 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851067 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851024 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12635888 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12684557 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851066 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851123 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30850953 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851043 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851119 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851056 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30850930 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12896164 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851103 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851032 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851063 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12885957 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851030 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30850971 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851062 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30850965 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30850981 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851001 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851026 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30850975 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851071 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30850969 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851073 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851042 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30850970 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30850983 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851044 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12789145 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851035 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851000 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12878192 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30850945 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851016 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30850995 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851088 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30850951 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30850973 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851090 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30850984 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851114 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30850986 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851003 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851011 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12789138 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851005 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851015 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30850959 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30850926 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851109 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851078 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30850944 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30850933 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30850979 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851100 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851046 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851097 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30850991 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851039 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851115 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851037 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851022 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851064 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851082 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851068 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851116 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851084 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851112 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851120 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851086 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30850949 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851021 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30850954 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851089 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851095 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851007 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30850964 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30850972 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851009 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30850937 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30850990 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851048 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851127 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30850940 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30850977 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30850934 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851051 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30850924 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851093 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851034 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851094 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12635885 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851054 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12635876 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30850942 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12988363 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851069 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30850967 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30851028 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Murine Genome U74v2 Set | 131092_PM_at | - | - |
| Murine Genome U74v2 Set | 131092_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 411205 | - | - |

D16Ertd519e
Genomic Map