Assay Details
| Gene Symbol: Gm29687 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK139823.1 | - | 3 - 4 | 538 | 76 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC328066 | LOC328066 | C920021A13 | -625.5kb | Anti-sense | Chr. 12 - 25316852 - 25471867 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 4930480M12 gene | 4930480M12Rik | 80.6kb | Anti-sense | Chr. 12 - 26211267 - 26240707 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm29687
Genomic Map