Assay Details
| Gene Symbol: Gm2464 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK140446.1 | - | 1 - 2 | 438 | 69 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 10745 | Gm10745 | ENSMUSG00000074696 | -1.6mb | Sense | Chr. 3 - 11823229 - 11844359 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 4930539N22 gene | 4930539N22Rik | 125.3kb | Sense | Chr. 3 - 13597139 - 13652865 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm2464
Genomic Map