Assay Details
| Gene Symbol: Slc1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK141069.1 | - | 1 - 2 | 321 | 74 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | peptidase domain containing associated with muscle regeneration 1 | Pamr1 |
5930437L24, E430002G05Rik, Ramp |
-141.3kb | Sense | Chr. 2 - 102550021 - 102643041 on NCBI Build GRCm38 |
| Downstream | CD44 antigen | Cd44 |
AU023126, AW121933, AW146109, HERMES, Ly-24, Pgp-1 |
22.4kb | Anti-sense | Chr. 2 - 102811141 - 102901669 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Slc1a2
Genomic Map