Assay Details
| Gene Symbol: A830039H05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK141989.1 | - | 1 | 3750 | 88 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | reticulon 3 | Rtn3 | RTN3-A1 | Sense | Chr. 19 - 7425895 - 7483291 on NCBI Build GRCm38 | |
| Downstream | atlastin GTPase 3 | Atl3 |
4633402C03Rik, 5730596K20Rik, AI465397, AW228836 |
24.4kb | Anti-sense | Chr. 19 - 7493816 - 7538609 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

A830039H05Rik
Genomic Map