Assay Details
Gene Symbol: Tmem207 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK142524.1 | - | 2 - 3 | 171 | 84 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | claudin 16 | Cldn16 |
PCLN1, claudin-16 |
-33.9kb | Anti-sense | Chr. 16 - 26463135 - 26482765 on NCBI Build GRCm38 |
Downstream | interleukin 1 receptor accessory protein | Il1rap |
6430709H04Rik, AI255955, AV239853, IL-1RAcP |
56.9kb | Anti-sense | Chr. 16 - 26581688 - 26730128 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() |
