| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10524081 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10524080 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10532312 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10524079 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10532310 |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0500009586.mm.1 |
TC0500001127.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500009586.mm.1 |
NR_040366.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500009586.mm.1 |
NR_040366.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500009588.mm.1 |
TC0500001127.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500009588.mm.1 |
NR_040366.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500009588.mm.1 |
NR_040366.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500009584.mm.1 |
TC0500001127.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500009584.mm.1 |
NR_040366.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500009584.mm.1 |
NR_040366.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500009585.mm.1 |
TC0500001127.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500009585.mm.1 |
NR_040366.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500009585.mm.1 |
NR_040366.1 |
Refseq |