Assay Details
| Gene Symbol: Gm8179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046039.1 | 1 - 2 | 368 | 92 | ||
| GenBank mRNA | AK143160.1 | - | 1 - 2 | 370 | 92 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 26578 |
Gm26578 | -133.5kb | Sense | Chr. 8 - 32009913 - 32018405 on NCBI Build GRCm38 | |
| Downstream | predicted gene 3985 | Gm3985 | 709.4kb | Anti-sense | Chr. 8 - 32888505 - 32950026 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm8179
Genomic Map