Assay Details
| Gene Symbol: Pbxip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146131.2 | NP_666243.1 | 11 | 3323 | 64 | |
| GenBank mRNA | AK047898.1 | - | 2 | 1168 | 64 | |
| AK081345.1 | - | 2 | 1168 | 64 | ||
| AK143270.1 | - | 1 | 956 | 64 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | pygopus 2 | Pygo2 | 1190004M21Rik | -13.7kb | Sense | Chr. 3 - 89430132 - 89435131 on NCBI Build GRCm38 |
| Downstream | phosphomevalonate kinase | Pmvk |
1110011E12Rik, 2900002L22Rik, AV001327, PMK, PMKA, PMKASE |
4.1kb | Sense | Chr. 3 - 89454546 - 89469014 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pbxip1
Genomic Map