Assay Details
| Gene Symbol: Gm1720 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046015.1 | 3 - 4 | 170 | 87 | ||
| GenBank mRNA | AK144520.1 | - | 3 - 4 | 171 | 87 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
EGF-like-domain, multiple 6 |
Egfl6 | Maeg | -77.7kb | Anti-sense | Chr. X - 166523007 - 166585716 on NCBI Build GRCm38 |
| Downstream | predicted gene 8817 | Gm8817 | EG667794 | 399.5kb | Anti-sense | Chr. X - 167063449 - 167070370 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC0X00001615.rn
Set Membership: |
|

Gm1720
Genomic Map