Assay Details
| Gene Symbol: Gm29680 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK144747.1 | - | 2 - 3 | 180 | 59 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transmembrane protein 167 | Tmem167 |
0610041E09Rik, 5730424F14Rik, AU041184, AW537806, ENSMUSG00000060669, Gm10085, Tmem167a |
-97.1kb | Anti-sense | Chr. 13 - 90089667 - 90114921 on NCBI Build GRCm38 |
| Downstream |
ATPase, H+ transporting, lysosomal accessory protein 1-like |
Atp6ap1l | EG435376 | 666.9kb | Sense | Chr. 13 - 90883439 - 90905355 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm29680
Genomic Map