Assay Details
| Gene Symbol: Gm29680 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK144747.1 | - | 5 - 6 | 516 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transmembrane protein 167 | Tmem167 |
0610041E09Rik, 5730424F14Rik, AU041184, AW537806, ENSMUSG00000060669, Gm10085, Tmem167a |
-64.3kb | Anti-sense | Chr. 13 - 90089667 - 90114921 on NCBI Build GRCm38 |
| Downstream |
ATPase, H+ transporting, lysosomal accessory protein 1-like |
Atp6ap1l | EG435376 | 700.6kb | Sense | Chr. 13 - 90883439 - 90905355 on NCBI Build GRCm38 |
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 495802 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 401617 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 669258 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 44184 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 110867 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 596468 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 539276 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 868799 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 462183 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 81079 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 133771 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P151032 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P151032 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07948448 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P151032 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212462 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212446 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212494 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212549 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212535 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212554 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212429 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212410 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212551 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212422 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212475 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212426 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10775595 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212314 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10550810 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212483 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212407 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212508 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212437 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212491 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212443 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212525 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212308 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212460 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212533 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212453 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10801027 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212480 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212403 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212325 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212326 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212315 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10585203 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10972549 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10708105 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212493 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212512 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212324 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10852952 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212417 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212537 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212498 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212501 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212310 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212412 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212528 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11096102 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212442 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212488 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11061924 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10536067 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212502 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212436 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212507 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10782568 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212503 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212434 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10972550 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212481 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212432 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10529038 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11096099 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10629827 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212458 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11113938 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212439 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212416 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212318 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10972547 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212322 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212304 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212321 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212499 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212496 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212511 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212477 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212555 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212447 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212552 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212532 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212486 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212520 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212414 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212408 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212504 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10585219 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212484 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212306 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212420 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212457 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212451 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212474 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212471 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212313 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212553 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212524 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212445 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212530 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212461 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10536069 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212455 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29212485 | - | - |

Gm29680
Genomic Map