Assay Details
| Gene Symbol: G730013B05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040380.1 | 1 - 2 | 135 | 69 | ||
| GenBank mRNA | AK144624.1 | - | 1 - 2 | 136 | 69 | |
| AK144852.1 | - | 1 - 2 | 77 | 69 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | bobby sox homolog (Drosophila) | Bbx |
5530401J07Rik, 5730403O13Rik |
-93.8kb | Anti-sense | Chr. 16 - 50191844 - 50432461 on NCBI Build GRCm38 |
| Downstream | coiled-coil domain containing 54 | Ccdc54 |
1700007N18Rik, AI644412, BB013989, NYD-SP17 |
52.3kb | Anti-sense | Chr. 16 - 50589859 - 50591154 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

G730013B05Rik
Genomic Map