Assay Details
| Gene Symbol: 2210406O10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK008838.1 | - | 1 - 2 | 298 | 74 | |
| AK144870.1 | - | 1 - 2 | 308 | 74 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | sortilin-related VPS10 domain containing receptor 2 | Sorcs2 |
N28137, mKIAA1329 |
-57.7kb | Sense | Chr. 5 - 36017180 - 36398167 on NCBI Build GRCm38 |
| Downstream |
GrpE-like 1, mitochondrial |
Grpel1 |
AA408748, mt-GrpE#1, mt-Grpel1 |
364.0 | Anti-sense | Chr. 5 - 36465185 - 36474077 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

2210406O10Rik
Genomic Map