Assay Details
| Gene Symbol: Gm805 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046081.1 | 1 - 2 | 104 | 68 | ||
| GenBank mRNA | AK144886.1 | - | 1 - 2 | 104 | 68 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | intraflagellar transport 43 | Ift43 |
1700019E19Rik, R75064 |
-6.9kb | Sense | Chr. 12 - 86082533 - 86162459 on NCBI Build GRCm38 |
| Downstream | G patch domain containing 2 like | Gpatch2l |
1700020O03Rik, 4732487G21Rik, A630090I05Rik, Kiaa1152 |
71.6kb | Sense | Chr. 12 - 86241838 - 86291793 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC0600001203.rn
Set Membership: |
|

Gm805
Genomic Map