Assay Details
| Gene Symbol: Tbx20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194263.2 | NP_919239.1 | 8 | 5087 | 104 | |
| GenBank mRNA | AK145503.1 | - | 1 | 1552 | 104 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | dpy-19-like 2 (C. elegans) | Dpy19l2 |
4932443J21Rik, Gm18376 |
-24.0kb | Sense | Chr. 9 - 24557047 - 24696941 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 29824 |
Gm29824 | 52.0kb | Anti-sense | Chr. 9 - 24775724 - 24795894 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tbx20
Genomic Map