Assay Details
| Gene Symbol: BC010981 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK145818.1 | - | 1 | 2247 | 93 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
ATPase, aminophospholipid transporter-like, class I, type 8A, member 2 |
Atp8a2 |
AI415030, Atpc1b, wl |
-26.9kb | Sense | Chr. 14 - 59647320 - 60197079 on NCBI Build GRCm38 |
| Downstream | microRNA 719 | Mir719 |
Mirn719, mmu-mir-719 |
2.0kb | Sense | Chr. 14 - 60228806 - 60228915 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

BC010981
Genomic Map