Assay Details
| Gene Symbol: E130317F20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK148204.1 | - | 1 - 2 | 136 | 96 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | polypyrimidine tract binding protein 1 | Ptbp1 |
AA407203, AL033359, HNRPI, PTB-1, PTB2, PTB3, PTB4, Ptb, pPTB |
Anti-sense | Chr. 10 - 79854432 - 79864435 on NCBI Build GRCm38 | |
| Downstream | lipid phosphate phosphatase-related protein type 3 | Lppr3 | Prg2 | 5.5kb | Sense | Chr. 10 - 79860475 - 79874634 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

E130317F20Rik
Genomic Map