Assay Details
| Gene Symbol: 1700052K11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027956.1 | 1 | 1874 | 100 | ||
| GenBank mRNA | AK148336.1 | - | 1 | 1749 | 100 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | methyltransferase like 2 | Mettl2 |
2810438F06Rik, AW493404, C130031G21Rik, D11Ertd768e, METL, PSENIP1 |
-37.9kb | Anti-sense | Chr. 11 - 105126425 - 105141146 on NCBI Build GRCm38 |
| Downstream |
mannose receptor, C type 2 |
Mrc2 |
Endo180, mKIAA0709, uPARAP |
111.3kb | Anti-sense | Chr. 11 - 105292576 - 105351146 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1700052K11Rik
Genomic Map