Assay Details
| Gene Symbol: Hnrnpf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK148509.1 | - | 168 | 90 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
guanine nucleotide binding protein (G protein), gamma 4 |
Gng4 | -34.6kb | Sense | Chr. 13 - 13784059 - 13827895 on NCBI Build GRCm38 | |
| Downstream |
UDP-GalNAc:betaGlcNAc beta 1,3-galactosaminyltransferase, polypeptide 2 |
B3galnt2 |
A930105D20Rik, C80633, D230016N13Rik |
86.4kb | Sense | Chr. 13 - 13954474 - 13999098 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Hnrnpf
Genomic Map