Assay Details
| Gene Symbol: Dpysl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136086.2 | NP_001129558.1 | 14 | 4454 | 102 | |
| NM_001291455.1 | NP_001278384.1 | 14 | 4704 | 102 | ||
| NM_009468.5 | NP_033494.1 | 14 | 4635 | 102 | ||
| GenBank mRNA | AK148950.1 | - | 1 | 954 | 102 | |
| AK149008.1 | - | 1 | 801 | 102 | ||
| AK149239.1 | - | 1 | 865 | 102 | ||
| Gene Symbol: 9430041P20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AV162270.2 | - | 1 | 265 | 102 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | serine/threonine kinase 32A | Stk32a |
A930015B13Rik, YANK1 |
-3.5kb | Anti-sense | Chr. 18 - 43207697 - 43317481 on NCBI Build GRCm38 |
| Downstream |
eukaryotic translation initiation factor 3, subunit J2 |
Eif3j2 |
ENSMUSG00000043424, Eif3j-2, Gm9781, eIF3 p35, eIF3j-B |
152.7kb | Sense | Chr. 18 - 43475418 - 43477836 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Dpysl3
Genomic Map