Assay Details
| Gene Symbol: Gm10431 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK154292.1 | - | 1 | 3750 | 73 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | G protein-coupled receptor 68 | Gpr68 |
BB131428, Ogr1 |
Sense | Chr. 12 - 100876682 - 100908198 on NCBI Build GRCm38 | |
| Downstream | coiled-coil domain containing 88C | Ccdc88c |
0610010D24Rik, AV000935, AW324073, Daple, mKIAA1509 |
12.1kb | Sense | Chr. 12 - 100911521 - 101028983 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm10431
Genomic Map