Assay Details
| Gene Symbol: Gm6093 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK161362.1 | - | 1 - 2 | 1242 | 68 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
serine (or cysteine) peptidase inhibitor, clade B, member 9g |
Serpinb9g |
1600002F03Rik, AW545779, NK21B |
-69.2kb | Anti-sense | Chr. 13 - 33484790 - 33496002 on NCBI Build GRCm38 |
| Downstream |
serine (or cysteine) peptidase inhibitor, clade B, member 6d |
Serpinb6d |
Gm11390, OTTMUSG00000000712, SPI3D |
92.5kb | Anti-sense | Chr. 13 - 33661405 - 33671585 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm6093
Genomic Map