Assay Details
| Gene Symbol: Zfp612 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175480.4 | NP_780689.3 | 5 | 4512 | 87 | |
| GenBank mRNA | AK162867.1 | - | 1 | 2246 | 87 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 8030455M16 gene | 8030455M16Rik | -30.6kb | Sense | Chr. 8 - 110039462 - 110059562 on NCBI Build GRCm38 | |
| Downstream |
predicted gene, 35543 |
Gm35543 | 21.0kb | Sense | Chr. 8 - 110113773 - 110121963 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Zfp612
Genomic Map