Assay Details
| Gene Symbol: I730030J21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045781.1 | 1 - 2 | 196 | 66 | ||
| NR_045782.1 | 1 - 2 | 196 | 66 | |||
| GenBank mRNA | AK146183.1 | - | 1 - 2 | 197 | 66 | |
| AK163102.1 | - | 1 - 2 | 180 | 66 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 6 | Galnt6 |
4632410F13, AW047994, GalNAc-T6 |
-552.0 | Sense | Chr. 15 - 100689915 - 100729929 on NCBI Build GRCm38 |
| Downstream |
solute carrier family 4 (anion exchanger), member 8 |
Slc4a8 |
AW493845, C230026C11, NDCBE, kNBC-3 |
29.0kb | Anti-sense | Chr. 15 - 100761700 - 100823971 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK163102.1 transcripts.
Set Membership: |
|

I730030J21Rik
Genomic Map