Assay Details
Gene Symbol: 6430550D23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK163252.1 | - | 339 | 79 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | centrosomal protein 250 | Cep250 |
AW490617, B230210E21Rik, Cep2, Inmp |
-3.5kb | Anti-sense | Chr. 2 - 155956272 - 155998900 on NCBI Build GRCm38 |
Downstream | ERGIC and golgi 3 | Ergic3 |
2310015B14Rik, AV318804, CGI-54, D2Ucla1, NY-BR-84, Sdbcag84 |
4.8kb | Anti-sense | Chr. 2 - 156008033 - 156018279 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
