Assay Details
| Gene Symbol: Eldr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK163834.1 | - | 1 | 55 | 100 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | epidermal growth factor receptor | Egfr |
9030024J15Rik, AI552599, Erbb, Errb1, Errp, Wa5, wa-2, wa2 |
-22.8kb | Anti-sense | Chr. 11 - 16752203 - 16913907 on NCBI Build GRCm38 |
| Downstream | F-box protein 48 | Fbxo48 | A630050E13Rik | 12.9kb | Anti-sense | Chr. 11 - 16951375 - 16954777 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Eldr
Genomic Map