Assay Details
| Gene Symbol: A230070E04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045897.1 | 2 - 3 | 1251 | 87 | ||
| GenBank mRNA | AK162870.1 | - | 2 - 3 | 1252 | 87 | |
| AK164995.1 | - | 1 - 2 | 2008 | 87 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
neurofilament, medium polypeptide |
Nefm |
NF-M, NF160, NF165, Nef3, Nfm |
Anti-sense | Chr. 14 - 68119545 - 68125004 on NCBI Build GRCm38 | |
| Downstream | a disintegrin and metallopeptidase domain 7 | Adam7 |
9230104J12Rik, EAP1, EAPI |
366.0kb | Anti-sense | Chr. 14 - 68497336 - 68533689 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

A230070E04Rik
Genomic Map