Assay Details
| Gene Symbol: Tox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK169546.1 | - | 1 - 2 | 62 | 65 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 11802 | Gm11802 | OTTMUSG00000004331 | -273.1kb | Anti-sense | Chr. 4 - 6700439 - 6717248 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 8430436N08 gene | 8430436N08Rik | 569.9kb | Anti-sense | Chr. 4 - 7560688 - 7573801 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Tox
Genomic Map