Assay Details
| Gene Symbol: Ankra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK171682.1 | - | 1 | 1944 | 102 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
UTP15, U3 small nucleolar ribonucleoprotein, homolog (yeast) |
Utp15 | AW544865 | -150.0 | Anti-sense | Chr. 13 - 98246845 - 98262992 on NCBI Build GRCm38 |
| Downstream | basic transcription factor 3 | Btf3 | 1700054E11Rik | 43.7kb | Anti-sense | Chr. 13 - 98309897 - 98317006 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ankra2
Genomic Map