Assay Details
| Gene Symbol: Slc25a51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK172225.1 | - | 1 | 554 | 82 | |
| BC026435.1 | - | 1 | 50 | 82 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | DDB1 and CUL4 associated factor 10 | Dcaf10 |
AA959934, Wdr32 |
-27.4kb | Anti-sense | Chr. 4 - 45341735 - 45379732 on NCBI Build GRCm38 |
| Downstream | src homology 2 domain-containing transforming protein B | Shb | BC028832 | 14.5kb | Sense | Chr. 4 - 45423276 - 45530828 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Slc25a51
Genomic Map