Assay Details
| Gene Symbol: Slc7a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK183612.1 | - | 1 - 2 | 71 | 83 | |
| AK189161.1 | - | 1 - 2 | 71 | 83 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 42049 |
Gm42049 | -13.8kb | Sense | Chr. 8 - 121881150 - 121882304 on NCBI Build GRCm38 | |
| Downstream | cDNA sequence BC048644 | BC048644 | 11.3kb | Anti-sense | Chr. 8 - 121907833 - 121918428 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Slc7a5
Genomic Map