Assay Details
| Gene Symbol: Acbd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK215959.1 | - | 2 - 3 | 202 | 98 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
phospholipase C, delta 3 |
Plcd3 |
2610205J15Rik, mKIAA1964 |
-9.6kb | Anti-sense | Chr. 11 - 103069236 - 103101658 on NCBI Build GRCm38 |
| Downstream | hexamethylene bis-acetamide inducible 1 | Hexim1 |
7330426E13Rik, CLP-1, Clp1, HIS1 |
4.6kb | Sense | Chr. 11 - 103116325 - 103119725 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Acbd4
Genomic Map