Assay Details
| Gene Symbol: Hivep3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY454346.1 | - | 1 - 2 | 74 | 53 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | guanylate cyclase activator 2b (retina) | Guca2b |
AV066530, Gcap2, Ugn |
-74.8kb | Anti-sense | Chr. 4 - 119656601 - 119658945 on NCBI Build GRCm38 |
| Downstream | endothelin 2 | Edn2 | VIC | 419.5kb | Sense | Chr. 4 - 120161424 - 120167360 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Hivep3
Genomic Map