Assay Details
| Gene Symbol: 2900008C10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045434.1 | 2 - 3 | 146 | 93 | ||
| GenBank mRNA | BC027704.1 | - | 2 - 3 | 146 | 93 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 4906 | Gm4906 | EG236663 | -808.3kb | Anti-sense | Chr. X - 11327822 - 11328151 on NCBI Build GRCm38 |
| Downstream | predicted gene 14635 | Gm14635 | OTTMUSG00000017536 | 202.3kb | Sense | Chr. X - 12339779 - 12354996 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

2900008C10Rik
Genomic Map