Assay Details
| Gene Symbol: Kcnj9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC038600.1 | - | 1 | 1364 | 73 | |
| BC048090.1 | - | 1 | 753 | 73 | ||
| BC069221.1 | - | 1 | 288 | 73 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
immunoglobulin superfamily, member 8 |
Igsf8 |
AA033172, AU041109, ESTM34, EWI-2, KCT-4, PGRL, R75384 |
-660.0 | Anti-sense | Chr. 1 - 172312407 - 172319841 on NCBI Build GRCm38 |
| Downstream |
potassium inwardly-rectifying channel, subfamily J, member 10 |
Kcnj10 |
BIR10, BIRK-1, Kir1.2, Kir4.1 |
19.7kb | Anti-sense | Chr. 1 - 172341210 - 172374085 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Kcnj9
Genomic Map